Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
The Thermo Scientific RadHalo FM Spectroscopic Area Monitor is designed for permanent and constant area monitoring. Able to withstand nearly all environmental conditions and emergency conditions. Battery backup and multiple reachback methods insure full operation even in emergency situations. Simple layout allows field service support and onsite calibrations. Antenna and satellite extension kits for long distance communication capability especially when RadHalo has no view of the sky.- Rapid dose rate response
- Immediate accurate identification
- Autonomous operation anywhere
- Durable design
- Secure transmissions
- Detector Fusion Capable
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Spectroscopic Detector: 3 x 3 in. NaI | |
RadHalo FM-NSF-R | |
Fixed with Neutron and fixed satellite, with Radio |
CLYC | |
Ethernet, Wi-Fi, 2.4 GHz long range radio and Cellular reachback Capabilty |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction