Learn More
SCS R3 Series 2-Layer Rubber Roll
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Dissipative rubber worksurface mat doesn't generate static charge and controls discharge from conductors placed on surface.
Supplier: SCS 770073
Description
- Dissipative Dual Layer Rubber Material
Provides a worksurface that does not generate a static charge and will control the discharge rate from all conductors (including ESD susceptible items) that are placed on the surface. - Color (1 x 106 to < 1 x 109 ohms Rtt) Top Layer
Meets ANSI/ESD S20.20 worksurface required limit and recommendation of ANSI/ESD S4.1 for contact with ESD susceptible items. Electricals will not drift for the life of the mat. - Black Side Bottom Layer
The dedicated conductive layer reduces the resistance to ground (Rtg) from the top of the surface to the groundable point ground and allows the mats to be used with ESD workstation monitors. Electricals will not drift for the life of the mat. - High Strength Nitrile Rubber Compound
Constructed to withstand abrasion, tearing and may be used in soldering applications with flux and other chemicals. - Non-Humidity Dependence
Provides consistent electrical performance regardless of ambient humidity. - Lot Traceability Information
Provides tracking information for lifespan of the product and to ensure quality control. - Lead-free RoHS 2 and REACH compliant
- Made in the United States of America
Specifications
Rubber | |
Meets ANSI/ESD S20.20 worksurface required limit and recommendation of ANSI/ESD S4.1 for contact with ESD susceptible items | |
50 ft. x 48 in. | |
121.92 cm |
15.24 m | |
Light Blue | |
48 in. | |
50 ft. |