Learn More
Electron Microscopy Sciences Aurion R-Gent Silver Enhance Kit 250 ML
Supplier: Electron Microscopy Sciences 25522
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
AURION R-Gent SE-EM is a high efficiency silver enhancement reagent for electron microscopy. It intensifies the gold particles by homogeneous deposition of metallic silver on the particle surface. The resulting electron dense signal is easily detected and is compatible with heavy metal counterstaining. The reagent has extremely delayed auto-nucleation and can be used under standard laboratory light conditions. It also has low viscosity, which is especially advantageous for pre-embedding immunogold applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.