Learn More
Brady™ Quik-Align™ No Water Picto Polyester Label
Easily apply outdoor-durable Quik-Align™ labels to identify sites, chemical containers or other points of emphasis where unusual reactivity with water could occur.
Supplier: Brady™ 175251
Description
- Card-mounted polyester film characters feature pressure-sensitive adhesive
- Flexible material conforms to most surfaces and holds up to harsh environments, abrasion, and repeated spills and cleanings
- Weathers up to 8 years outdoors, even longer indoors, and withstands wide temperature variations (-40°F to +230°F)
Specifications
Laminated Polyester | |
Permanent Acrylic | |
3 in. | |
85.73 mm | |
-40°F to +230°F | |
Black on White |
10 Labels/Pk. | |
Facility Identification, Hazard Communication Identification, Safety Identification | |
3.375 in. | |
-40°C to +110°C | |
W |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov