Learn More
Zymo Research Corporation Quick SARS-CoV-2 Multiplex Kit, 1000 rxns
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Zymo Research Corporation R301310K
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
![Request Bulk Order](/content/dam/fishersci/glyphs/15395_Quote_Bulk_EN.jpg)
Results are for the identification of SARS-CoV-2 RNA. The SARS-CoV-2 RNA is generally detectable in upper and lower respiratory specimens, and oral fluids during the acute phaseof infection. Positive results are indicative of the presence of SARS-CoV-2 RNA. Positive results do not rule out bacterial infection or co-infection with other viruses. The agent detected may not be the definite cause of disease.Negative results do not preclude SARS-CoV-2 infection and should not be the sole basis of a patient treatment/management or public health decision.The Quick SARS-CoV-2 Multiplex Kit is intended for use by personnel specifically instructed and trained in the techniques of real-time RT-PCR assays.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.