Learn More
Electron Microscopy Sciences R 0.6/1 Holey Carbon Films on Grids, Grid: Gold. Mesh: 400., 25/PK
Supplier: Electron Microscopy Sciences Q420AR06
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
QUANTIFOIL is a perforated support foil with pre-defined hole size, shape and arrangement that has advantages in electron microscopy (EM) or low-energy electron point source (LEEPS) when compared with conventional holey film. QUANTIFOIL with circular holes is used in cryo-electron tomographic reconstruction. The roundness of the holes is advantageous with respect to the formation of an ice layer of constant thickness. Hole size is 0.6 µm. Space between holes is 1 µm. Center to center is 1.6 µm (hole size may be as large as 1 µm). Smaller holes are designed for magnifications of 50,000x or higher. Grid: Gold. Mesh: 400 .
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.