Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Purifier™ Logic™+ Class II A2 Biosafety Cabinets, 5 ft. Width, International Models
SDP

Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality

Supplier:  Labconco™ 302580141

Request Bulk Order

 View more versions of this product

Catalog No. 30-258-0141


Add to Cart

Alert:

This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.


Description

Description

All models feature:

  • Electronically Commutated Motor (ECM)
  • Constant Airflow Profile (CAP) airflow monitoring system
  • Intrinsically-safe negative pressure design
  • Air-Wave™ Entry System
  • Contain-Air™ Negative Pressure Channel
  • Supply and exhaust 99.99% efficient HEPA filters. 99.999% efficient ULPA filters available
  • Interior-mounted, line-of-sight color LCD with MyLogic OS. Displays filter life, status messages for alarm conditions and alerts. 8 languages available
  • Bright, 90-150 footcandle, glare-free LED lighting located outside the contaminated work area
  • Fully-closing, clear 1/4 in. tempered safety glass sash
  • Curved stainless steel inlet grille with Reserve-Air Secondary Airflow Slots
  • Two electrical duplex receptacles. Flush-mounted-stainless steel with dampened hinges. Single outlets on 230 V models. One outlet on each side, with ground fault interruption (115 V)
  • Smart-Start™ System user-programmable start up and shut down
  • Night-Smart™ System reduces blower speed when sash is closed
  • Leak-tight stainless steel interior and powder-coated steel exterior
  • Removable towel catch located under work surface
  • Code-activated electronic security lock
  • 22.6 in. (57 cm) max. sash opening and 27 in. (69 cm) viewing height
  • 10° Angled sash with counterbalanced, easy lift mechanism
  • ADA-Compliant touchpad control on right side post
  • Noise level <63 dBA
  • Heat load 990 BTU/hour (4 ft. models)
  • Removable, seamless type 304 stainless steel, dished work surface with lift out knobs
  • Nominal inflow velocity of 105 fpm (0.5 m/sec)
  • Nominal downflow velocity of 55 fpm (0.3 m/sec)
  • Approximately 70% air recirculation
  • Overall depth x height: 31.2 x 61.7 in. (79.2 x 156.7 cm)
  • 10 ft (3 m) power cord with plug
  • Built-in timer and digital clock
  • Five year warranty
  • NSF International listed
  • Optional NSF-approved non-welded telescoping base stand included on some models
  • Optional Accessory Package (254 nm UV lamp, ADA-compliant service fixture(s) with ball-type valve(s), and NSF-Approved Vacu-Pass™ Cord & Cable Portal) included on some models
Specifications

Specifications

NSF, CE Marked, ADA, ISO 5 Conditions per ISO 14644-1 and 2
153.7 cm
Purifier Logic+
2
340 W
89.2 to 95.2 in.
66.3 in.
168.402 cm
LED
8 in.
60.5 in.
50/60 Hz
79.248 cm
230 V
Stainless Steel
Type A2 Biosafety Cabinet
<63 dBA
China, Australia (230 V)
2
20.3 cm
0.159 to 0.174 m3sec.
Cabinet, Base Stand
226.6 to 241.8 cm
5 Years
168.402 cm
31.2 in.
230 V, 50/60 Hz, 8 A
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit