Learn More
Biolegend Purified anti-TFAP2A [W20235D]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: WB, ICC, ICFC, IHC-P; Size: 25 μg
Supplier: Biolegend 634351
Purified anti-TFAP2A [W20235D]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: WB, ICC, ICFC, IHC-P; Size: 25 μg