Learn More
Gentex™ PureFlo 3000 Pre-Filter
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Pre filter (50-pack) for use with the PureFlo 3000 PAPR.
Supplier: Gentex™ PF300004005
This item is not returnable.
View return policy
Description
- Pre filter for use with the PureFlo 3000 PAPR.
- To provide respiratory protection against dusts, mists and aerosols.
Specifications
PARTICULATE | |
Prefilter | |
Yes | |
50/Pk. | |
Non-sterile |
White | |
PureFlo 3000 PAPR | |
PureFlo 3000 PAPR | |
Yes |