Learn More
Electron Microscopy Sciences PTFE Beaker Cap, 5 ml
Supplier: Electron Microscopy Sciences 60938C
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Is statically molded from virgin "PTFE" and machine finished to give a smooth surface for ease of cleaning and to eliminate contamination. Supplied with a pouring spout and they are virtually immune to chemical attack. Working temperature range is from cryogenic to 260 degree C (500 degree F). Ideal for use in wafer etching.
Capacity: 5 ml
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.