Learn More
World Precision Instrument Patch Clamp Glass-PG52165-4
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: World Precision Instrument PG521654
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
PG52151-4 and PG52165-4 are prepared from Schott #8250 glass (equivalent to Corning #7052), one of the most widely used patch clamping glasses. This is a specially formulated borosilicate glass with a softening temperature that is 110°C lower than regular borosilicate glass (Corning 7740, or Pyrex). It has excellent sealing properties for most cells. Electrical properties are also very good.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.