Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ PTC 209
Bmi-1 inhibitor; antitumor
$343.36 - $1438.40
Specifications
Purity | >98% |
---|---|
Product Type | PTC 209 |
Molecular Weight (g/mol) | 495.19 |
Inhibitors | PTC 209 |
Description
Bmi-1 inhibitor (IC50 ∽ 0.5 μM); irreversibly impairs colorectal cancer-initiating cell (CIC) growth. Reduces tumor growth in CIC xenograft assays and results in reduced potential of colorectal cancer cells to initiate tumors in vivo.Specifications
>98% | |
PTC 209 | |
495.19 | |
PTC 209 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title