Learn More
ERA WATERS CORP PotableWatR Coliform MicrobE
SDP
Supplier:  ERA WATERS CORP 080B
Encompass
Includes 10 lyophilized standards with at least 1 total and fecal positive, 1 total positive and fecal negative, 1 total and fecal negative, and 1 blank. The standards are applicable to all SDWA promulgated methods–MF, MPN, presence/absence and ONPG-MUG.
Catalog No. NC1174809