Learn More
Applied Physics PSL SPHERES, 50 NM, 15ML BOTTL
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Applied Physics AP3050A15M
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
PSL Spheres, 50 nm, 15ml bottle volume, Polystyrene Latex Beads, NIST Traceable, Size Calibration Certificate included
This item is not returnable.
View return policy