Learn More
Medchemexpress LLC HY-15005 100mg , PSI-7977 GS 7977;sofosbuvir CAS:1190307-88-0 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY15005100MG
Encompass_Preferred
Medchemexpress, HY-15005 100mg PSI-7977 GS 7977;sofosbuvir CAS:1190307-88-0 Formula:C22H29FN3O9P EC50: 92±5 nM (HCV) Purity:98% PSI-7977 is an active inhibitor of HCV RNA replication in the HCV replicon assay, demonstrates potent anti-hepatitis C virus ( HCV ) activity. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2705
Due to product restrictions, please to purchase or view availability for this product.