Learn More
Electron Microscopy Sciences PS4R with NPL Certificate
Supplier: Electron Microscopy Sciences 680474RNP
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
These stage micrometers are available with a calibration certificate that provides traceability for the precise calibration and confirmation of accuracy of optical measuring instruments, necessary under ISO provisions. The glass discs are mounted in stainless steel slides with engraved serial numbers. Each slide is supplied in a polished wooden storage case. Reflected light stage micrometer with 0.1" scale subdivided to 0.001". Calibrated by the National Physical Laboratory by taking measurements at selected points on the scale.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.