Learn More
Invitrogen™ Proteinase K (50 μg/μL)
This Proteinase K is used in QuantiGene assays and is one of two components in the QuantiGene Sample Processing kits.
$81.25 - $1054.00
Specifications
Content And Storage | Store in freezer (-5°C to -30°C). |
---|---|
Shipping Condition | Blue Ice |
Form | Liquid |
Product Type | Proteinase K |
Description
Proteinase K digests proteins such as ribonucleases that could interfere with the quantitation of RNA using QuantiGene assays.
Specifications
Store in freezer (-5°C to -30°C). | |
Blue Ice | |
Liquid | |
Proteinase K |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.