Learn More
Medchemexpress LLC HY-111846 5mg Medchemexpress, PROTAC ERα Degrader-2 CAS:1351169-29-3 Purity:>98%
Supplier: Medchemexpress LLC HY1118465MG
Medchemexpress, HY-111846 5mg PROTAC ERα Degrader-2 CAS:1351169-29-3 PROTAC ERα Degrader-2 comprises a cIAP1 ligand binding group, a linker and an estrogen receptor α (ERα) binding group. PROTAC ERα Degrader-2 is an ERα degrader. Maximal ERα degradation at 30 μM concentration in human mammary tumor MCF7 cells. Degradation inducers based on cIAP1 are called specific and non-genetic IAP-dependent protein erasers (SNIPERs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.