Learn More
Enzo Life Sciences Progranulin (human), (recombinant) (50µg)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Enzo Life Sciences ALX2015020050
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Alternative name: Proepithelin, PEPI, PC cell derived growth factor. Endotoxin: <0.01EU/µg protein (LAL-test). Purity: ≥95% (SDS-PAGE). MW: ~90-95kDa. Formulation: Lyophilized from a 0.2 μm filtered solution in PBS. Source: Produced in mouse myeloma cell line NS0. Human progranulin (aa 18-593) is fused at the C-terminus to a 6-His tag. Long Term Storage: -20°C. Use/Stability: Working aliquots are stable for up to 3 months when stored at -20°C. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.