Learn More
Cole-Parmer Oakton Conductivity Probe, 2-cell, K = 0.1, Epoxy Body/Pt bands, 2.5-ft Cable
Supplier: Cole-Parmer 3560872
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Keep your multiparameter meters up and running with replacement conductivity probes.