Learn More
Haztech Systems, Inc HW5111 Pro-8, 8 strip (Anthrax, Ricin, Botulinum, SEB, Plague, Tularemia, Brucella & Orthopox
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Haztech Systems, Inc HW5111
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
HW5111 Pro-8, 8 strip (Anthrax, Ricin, Botulinum, SEB, Plague, Tularemia, Brucella & Orthopox) Screening device designed to quickly evaluate a suspicious powder or liquid in the field or in the laboratory. Used in conjunction with Hazcat's protein test. Accurately detects minute amounts of specific biological threat agents.