Learn More
Gilson™ Pro Pipette Service for Multichannel Pipettes
Gilson™ Pro Pipette Service offers quality service, preventative maintenance, and calibration to ensure pipettes lasts for years to come.
Supplier: Gilson™ SVC101131
Description
- Visual and Functional Inspection
- Internal and External Cleaning
- Leak Test
- Gravimetric Check
- Preventative Maintenance
- Re-grease as needed
- Pipette Label
- Mail-In or Onsite Service Available
- Calibration and Detailed Certificate
- ISO 17025 Calibration Available
- 5 day turn around time
Specifications
8-channel Pipettes | |
Data collected before and after repair | |
Pipette Pro PM and Calibration Service |
ISO 17025 Accredited Service | |
5 Days | |
4 data points collected at 2 volumes |