Learn More
STEMCELL Technologies Human Peripheral Blood Plasma, Fresh, Size: 10 mL
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: STEMCELL Technologies 2000150
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Primary human plasma has been isolated from peripheral blood (PB) using centrifugation. PB was collected using acid-citrate-dextrose solution A (ACDA). Human plasma is the liquid component of blood containing water, salts, lipids, hormones, proteins (including albumin), immunoglobulins, clotting factors, and fibrinogen. It does not contain the cellular components of blood—the erythrocytes and the buffy coat composed of leukocytes and platelets. Plasma, from a single donor, was obtained using Institutional Review Board (IRB)-approved consent forms and protocols.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.Browse our frequently asked questions (FAQs) on primary cells. (https://www.stemcell.com/technical-resources/educational-materials/tech-tips/primary-cells-faq.html)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.