Learn More
STEMCELL Technologies Human Peripheral Blood Serum, Fresh, Size: 5 mL
SDP
Supplier:  STEMCELL Technologies 2000158
Encompass_Preferred
Primary human serum has been isolated from normal donor peripheral blood (PB) after coagulation and removal of the fibrin clot and blood cells. Human serum is the fluid portion of blood that is left after clotting has removed clotting factors/fibrinogen and cellular components. Serum contains water, proteins, electrolytes, antibodies, antigens, hormones, and any exogenous substances.Serum, from a single donor, was obtained using Institutional Review Board (IRB)-approved consent forms and protocols.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.Browse our frequently asked questions (FAQs) on primary cells. (https://www.stemcell.com/technical-resources/educational-materials/tech-tips/primary-cells-faq.html)
Catalog No. 50-206-9406
Explore available promotions