Learn More
Hach Company Presence-Absence (P-A) Broth, Dehydrated 500 g
Supplier: Hach Company 2280934
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Used to detect pollution in treated or potable water; described in the 16th ed., Standard Methods.