Learn More
Andwin Scientific PRE-MOISTENED TOWELETTES
Supplier: Andwin Scientific 52848078
The Sight Savers Premoistened Lens Cleaning Tissues are ideal for sanitizing all glass and plastic lenses, such as computer screens, mirrors, safety glasses, and face shields. This product is treated with surfactants in order to efficiently perform its tasks. It is conveniently packaged and sold in a cardboard dispenser box to permit quick and easy dispensing of large 12.7x20.3cm (5x8") folded, premoistened tissues.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.