Learn More
TSI Incorporated PORTACOUNT FIT TEST MODEL 8038
Supplier:  TSI Incorporated 8038
Encompass
Request Bulk Order
No other respirator fit tester can quantitatively fit test all types of respirators-gas masks, SCBAs, respirators, even N95, P1 and P2 disposable (filtering-facepiece) respirators. The PortaCount Pro+ Respirator Fit Tester 8038 eliminates the guesswork associated with tedious and error-prone qualitative fit test methods. If the mask requires fit testing, count on the PortaCount Pro+ Fit Tester to provide the fastest, easiest and best HSE (UK) and OSHA (US)-compliant fit test method.
Catalog No. NC0815121
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.