Learn More
Cole-Parmer Airfiltronix 200A HS3000A1 Portable Fume Extractor, Single Hose; 115 VAC
Supplier: Cole-Parmer 3370543
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Inexpensive solution for capturing hazardous fumes and particles. Ideal for use in work spaces too small for conventional fume hoods. Can be used on tabletop or wall mounted. Flexible 5 ft hose can easily be placed at the source. Accommodates a variety of filters to meet most applications.