Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Heathrow Scientific™ Polystyrene Sterile Reagent Reservoir
Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures.
$224.00 - $296.00
Specifications
Color | White |
---|---|
Material | Polystyrene |
Sterility | Sterile |
Packaging | 5 Wrapped |
Quantity | 5/Bag, 40 Bags/Pk |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
03-449-258
|
Heathrow Scientific™
120640 |
25 mL |
Pack of 200 for $224.00
|
![]() |
|||||
03-449-261
|
Heathrow Scientific™
120643 |
50 mL |
Pack of 200 for $281.50
|
![]() |
|||||
03-449-265
|
Heathrow Scientific™
120647 |
100 mL |
Pack of 200 for $296.00
|
![]() |
|||||
Description
- Trough design with a v shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
- Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
- Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
- Designed to stay flat on the benchtop and not warp
- Sterile, sealed in bags and electron beam sterilized
- DNase, RNase and Pyrogen free
- Sealed in easy-to-open bags
- Quality certificate available on request
- Disposable
Specifications
White | |
Sterile | |
5/Bag, 40 Bags/Pk | |
1 | |
Convoluted |
Polystyrene | |
5 Wrapped | |
No | |
Single-use |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title