Learn More
Fisherbrand™ Polystyrene Antistatic Weighing Dish PROMO
Disposable dishes are ideal for weighing, dispensing, or storing applications.
Catalog No. 08-732-113PL
Explore available promotions
Add to Cart
Description
Description
Ideal for weighing static-affected materials.
  • Resist dilute acids, aqueous solutions, alcohols, and bases
  • Flat bottoms resist tipping
  • Made from 12 mil thick polystyrene - 20% thicker than most dishes on the market today—for better strength, stability, and handling ease
  • Pour boats feature a molded spout for convenient pouring and dispensing
Specifications
Specifications
Flat
Natural
Polystyrene
Weighing Dish
12 mil
3.5 in.
3.5 in.
25.4 mm
61 mm
100 mL
Single-use
Boat
Ideal for weighing, dispensing or storing applications
3.5 x 3.5 x 1 in. (89 x 89 x 25.4 mm)
89 mm
89 mm
1 in.
61 mm (Inner Base)
Product Suggestions
Product Suggestions