Learn More
Waters Corp Polyethylene Oxide Kit
Supplier:  Waters Corp WAT011572
Encompass_Preferred
500 mg/vial Polyethylene oxide kit for aqueous MW testing: 24,000, 79,000, 160,000, 340,000, 570,000, and 850,000.
Catalog No. 50-785-010
Explore available promotions