Learn More
Waters Corp Polyethylene Oxide Standard 340,000
Supplier:  Waters Corp WAT011582
Encompass_Preferred
An individual GPC standard (Gel Permeation Chromatography standards) for molecular weight determination. Polyethylene Oxide Standard at 340,000 Da
Catalog No. 50-785-015
Explore available promotions