Learn More
Monarch Instrument Pocket Laser Tach 200, Portable Tachometer
Supplier: Monarch Instrument 6125010
Tachometer; Pocket Laser 200; Includes: tachometer/timer/totalizer with N.I.S.T. traceable certificate of calibration, and 12 in. of Reflective Tape; Operates up to 25 ft.; Accepts Remote Sensors; Dim: 6.92H times; 2.4W times; 1.6 in. D