Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Verax Biomedical™ Platelet PGDprime™ Test
Rapid test for the detection of Gram-positive and Gram-negative bacterial contamination in platelets. Allows extension of platelet dating from 5 to 7 days when testing apheresis platelets in plasma.
$879.10 - $4395.50
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-051-017
|
Verax Biomedical™
PRM20 |
20 Tests |
Pack of 20 for $879.10
|
|
|||||
23-051-010
|
Verax Biomedical™
PRM100 |
100 Tests |
Pack of 100 for $4,395.50
|
|
|||||
Description
- Easy to perform qualitative test
- Detects both aerobes and anaerobes
- Observed specificity: 100% / LCL: 99.9%
- 150 μL sample size
- No centrifugation or precision pipetting required
- Includes reagents and disposables required to perform the test
- Ready to use liquid reagents in dropper bottles
- Colored line in read window indicates a reactive result
- Built in procedural control verifies assay validity
- FDA cleared for use with: Leukoreduced apheresis platelets in plasma or PAS-C, polls of up to 6 pre-storage or post-storage pooled WBD platelets and individual WBD platelets
- CLIA Complexitiy: Moderate
- Ready to use liquid reagents in dropper bottles
- Colored line in read window indicates a reactive result
Specifications
No | |
Box |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title