Learn More
Raven Environmental Products Coretaker Sludge Sampler 24ft
Supplier: Raven Environmental Products B1010424
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Core sampler for measuring and viewing settled sludge and solids. CORETAKER has durable 1.25 in. OD clear polycarbonate tubes with black ABS machined Slip Lock Coupler and automatic check valve. No threaded fittings that break. Fittings are CNC machined for precision fit. Internal O-Rings prevent leaks. Thanks to a foam lanyard the entire unit will float if dropped in a tank. Comes in 8 ft. sections to minimize the number of couplers. Commonly used in wastewater treatment plants to monitor activated sludge blanket levels. Retrieve samples and spin in centrifuge to monitor Percent Solids for Waste Activated Sludge (WAS). Also used in onsite wastewater and septic systems. Packaged one unit per box. Exclusive Lifetime Break-Free Guarantee. Made in USA.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.