Learn More
TARGETMOL CHEMICALS INC PKUMDL-LC-101-D04 5MG
Supplier: TARGETMOL CHEMICALS INC T36935
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. PKUMDL-LC-101-D04 is a potent allosteric activator of glutathione peroxidase 4 (GPX4) a regulator of iron death. GPX4 activity was increased to 150% of control levels when used at a concentration of 20 uM in a cell-free assay and at a concentration of 61 uM in wild-type but not Gpx4-/- mouse embryonic fibroblast (MEF) extracts.PKUMDL-LC-101-D04 (200 uM) reduced cholesterol peroxide-induced MEF death. purity: 98%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.