Learn More
Bellco Glass, Inc. The Bellco Pipette Plugger is designed for plugging a full range of standardized cotton plugging pipettes. This unit has three speeds and can be adjusted quickly and easily to plug 40, 50 or 60 pipettes per minute. The unit will produce uniform shape cott
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Bellco Glass, Inc. 775011220
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
![Request Bulk Order](/content/dam/fishersci/glyphs/15395_Quote_Bulk_EN.jpg)
The Bellco Pipette Plugger is designed for plugging a full range of standardized cotton plugging pipettes. This unit has three speeds and can be adjusted quickly and easily to plug 40, 50 or 60 pipettes per minute. The unit will produce uniform shape cott