Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Order Info
- INJECTION KEY
- Plain: Circulatory system not injected with colors.
- Single: Arteries injected with red latex.
- Double: Arteries red, veins blue.
- Triple: Arteries red, veins blue, and hepatic portal veins yellow.
Specifications
Fetal Pig | |
Plain | |
27.94 to 33.02 cm |
Fisher-Free™ | |
11 to 13 in. | |
Ea. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction