Learn More
Echelon Biosciences Research Labs PI3-Kinase Activity Fluorescence Polarization Assay
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Echelon Biosciences Research Labs K1100EAKIT
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
The PI3-Kinase Activity Fluorescence Polarization Assay determines PI3-K activity through quantification of its product, PI(3,4,5)P3 (PIP3).Sample Type: in vitro determination of PI3-K activity in cancer tissues and cell lines and compound screening for drug discoverySample Volume: 2.5-5.0 L enzyme / inhibitorAssay Range: 0.0625 M to 4.0 M This item is non returnable
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.