Learn More
Mabtech Inc PepPool EBV CD4 and CD8 human
Supplier: Mabtech Inc 36411
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
PepPool EBV CD4 and CD8 humanPeptide pool with 100 peptides, 25 ug of each peptide. Supplied in two vials (lyophilized).