Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ PELTOR™ FLX2 Cable for CH-3 Listen Only Headset
Click to view available options
Model:
FLX2-202
FLX2-203
FLX2-204
FLX2-205
FLX2-208
For Use With (Equipment):
1/4 in. stereo plug
2.5 mm mono plug
3.5 mm mono plug
3.5 mm stereo plug
3.5 mm stereo threaded plug
Description
- For use with CH-3 Listen-Only Headsets
- Simple and reliable connection to a wide variety of handheld devices.
- Coiled polyurethane cable
- Aramid fiber reinforced
- Robust and highly flexible
Compliance
3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Color | Black |
For Use With (Equipment) | 2.5 mm mono plug |
For Use With (Application) | Enables connection to listen-only CH-3 Headsets with Hytera, Icom, Kenwood or Motorola radios |
Model | FLX2-203 |
Pairs per Package | 1 Ea. |
Product Type | FLX2 Cable |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction