Learn More
Premier Lab Supply Pellet Film, 32mm Diameter – PN# PF-32-500
Supplier: Premier Lab Supply PF32500
Mylar 12.0μm Pellet Film Pre-Cut Circles interleaved with PREMIER Kraft Paper (PN# PF-32-500) – Diameter 1.26” (32mm) – 500 per box.
This item is not returnable.
View return policy