Learn More
SCIENION US INC PDC 70 PIEZO DISP CAP, P-2030
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: SCIENION US INC P2030
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
PDC 70 Piezo Dispense CapillaryS5,11,SX,DW,S3 (4x manifold)Fixed drop volume between 300-360 pl
This item is not returnable.
View return policy