Learn More
Cell Biolabs Inc pCMV-VSVG Envelope Vector, 10 µg
Supplier: Cell Biolabs Inc RV110
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
pCMV-VSV-G Envelope Vector can be used in co-transfections with other packaging plasmids to make recombinant lentivirus or retrovirus.