Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ PALB2/CEN16q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
$2310.00
Specifications
Content And Storage | Store at 4°C in the dark. |
---|---|
Description | DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
Format | Tube |
For Use With (Application) | FISH-Ce |
Gene ID (Entrez) | 79728 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-955-634
|
Abnova™
FG0182 |
100 μL |
Each of 1 for $2,310.00
|
|
|||||
Specifications
Store at 4°C in the dark. | |
Tube | |
79728 | |
Liquid |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe | |
FISH-Ce | |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title