Learn More
Cell Biolabs Inc pAAV-RC4 Vector, 10 µg
Supplier:  Cell Biolabs Inc VPK424
Encompass_Preferred
Cell Biolabs' pAAV-RC4 Vector contains the rep and cap genes required to generated recombinant AAV of serotype 4. Co-transfect this vector with other packaging plasmids and an expression vector into 293 cells for AAV-4 packaging.
Catalog No. NC1523141
Explore available promotions