Learn More
Electron Microscopy Sciences Veco Oyster Hexagonal DH75/300, 75/300, Ni
Supplier: Electron Microscopy Sciences D753HNI
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Nickel
Pitch: 333/ µm
Hole: 293/ µm
Bar: 50/ µm