Learn More
Chemglass Life Sciences 1.7 Cu. Ft. Oven, Vacuum, 230V
SDP
Supplier:  Chemglass Life Sciences CLS3450217
Encompass_Preferred
Request Bulk Order
1.7 Cu. Ft. Oven, Vacuum, 230V. The Vacuum ovens are durably constructed with seamless welds & strong bracing. All of the vacuum ovens are built with a stainless steel chamber for exceptional durability. Ovens incorporate solid brass vacuum valves w/ PTFE seats to prevent leaks. The double plenum design exceeds CE safety requirements resulting in a cool outer surface. The doors on these units have positive latch handles with spring-loaded glass to facilitate a good vacuum seal without hinge binds that shorten the gasket life. A selection of gaskets (for specific applications) & a small footprint increase the versatility of these ovens. The unique cross-flow ventilation forces inert gas to fill the entire chamber. Users can choose from 3/8 inch orifices or a KF25 fitting to withstand heavy use, minimize draw-down time and achieve low vacuum levels. We offer a full line of gaskets to suit applications involving high temperatures, acids, or solvents. Ovens have a 24 month limited warranty
Catalog No. 50-143-536
Explore available promotions