Learn More
Ortho-Clinical Diagnostics™ VITROS™ Controls: Anti-HBc TOTAL
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ortho-Clinical Diagnostics™ 6801430
This item is not returnable.
View return policy
Specifications
VITROS™ ECi immunodiagnostic system | |
Anti-HBc Control | |
Qualitative detection of total Antibody (IgG and IgM) to Hepatitis B core Antigen (total Anti-HBc) | |
1 mL |