Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ortho-Clinical Diagnostics™ VITROS™ Calibrators: Rubella IGG
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ortho-Clinical Diagnostics™ 8209074
This item is not returnable.
View return policy
Specifications
2°C to 8°C | |
Human Serum and Plasma (Heparin, EDTA or Sodium Citrate) | |
0.85mL Rubella IgG calibrators 1, 2 and 3 (Human Plasma with antimicrobial agent), lot calibration card, protocol card, 24 calibrator bar code labels (8 for each calibrator) | |
3 |
VITROS™ ECi/ECiQ immunodiagnostic system, 3600 immunodiagnostic system, 5600 integrated system | |
Rubella IGG Calibrator | |
Quantitative measurement of Rubella IgG | |
1 Set/Pk. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction